Ethyl glycolate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate
M-Clarity™ quality level = MQ100
364843-25G
364843-5G
SIAL364843-25GEA
4810
CZK
SIAL364843-25G
SIAL364843-5G
Ethyl glycolate, Sigma-Aldrich®
Ethyl glycolate
Vzorec:
C₄H₈O₃ MW: 104,11 g/mol |
MDL Number:
MFCD00021970 CAS číslo: 623-50-7 EINECS: 210-798-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...