Tisk…

Ethyl glycolate ≥95%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate

41475.09 41475.18
41475.09EA 2100 CZK
41475.09 41475.18
Ethyl glycolate ≥95%
Ethyl glycolate

Purity: 95% CAS No.: 623-50-7 Molecular Formula: C4H8O3 Formula Weight: 104.11 Flash Point: 61°(143°F) Physical Form: Liquid Density: 1.10 Refractive Index: 1.4190 Appearance: Colourless Boiling Point: 158-159° MDL No.: MFCD00021970

Vzorec: C₄H₈O₃
MW: 104,11 g/mol
Bod varu: 158…159 °C
Hustota: 1,10
Bod vzplanutí: 61 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021970
CAS číslo: 623-50-7
EINECS: 210-798-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR