Tisk…

N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: t-boc-N-EDA

L19947.14 L19947.06
L19947.14EA 10680 CZK
L19947.14 L19947.06
N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol
N-Boc-ethylenediamine

Purity: 98% CAS No.: 57260-73-8 Molecular Formula: (CH3)3COCONHCH2CH2NH2 Formula Weight: 160.22 Flash Point: 110°(230°F) Refractive Index: 1.4570 Boiling Point: 72°/0.1mm Beilstein Reference: 1932330 UN No.: 2735 MDL No.: MFCD00191871

Vzorec: C₇H₁₆N₂O₂
MW: 160,22 g/mol
Bod varu: 72 °C (0,1 mmHg)
Hustota: 1,016
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191871
CAS číslo: 57260-73-8
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR