Tisk…

N-Boc-N-methylethylenediamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-Boc-(2-Aminoethyl)-N-methylamine, N-Methylethane-1,2-diamine, N-BOC protected

15567-1ML 15567-5ML
SIAL15567-1MLEA 12350 CZK
SIAL15567-1ML SIAL15567-5ML
N-Boc-N-methylethylenediamine, Sigma-Aldrich®
N-Boc-N-methylethylenediamine
Vzorec: C₈H₁₈N₂O₂
MW: 174,24 g/mol
MDL Number: MFCD01317789
CAS číslo: 121492-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR