Tisk…

Trimethylsilyl acetate (acetoxytrimethylsilane), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trimethylsilyl acetate

M-Clarity™ quality level = MQ100

284459-100G 284459-25G
SIAL284459-100GEA 4130 CZK
SIAL284459-100G SIAL284459-25G
Trimethylsilyl acetate (acetoxytrimethylsilane), Sigma-Aldrich®
Trimethylsilyl acetate (acetoxytrimethylsilane)
Vzorec: C₅H₁₂O₂Si
MW: 132,24 g/mol
MDL Number: MFCD00008695
CAS číslo: 2754-27-0
EINECS: 220-404-2
UN: 3272
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR