Tisk…

4'-Cyanoacetanilide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-(4-Cyanophenyl)acetamide

A14941.06 A14941.14
A14941.06EA 1010 CZK
A14941.06 A14941.14
4'-Cyanoacetanilide ≥98%
4'-Cyanoacetanilide

Purity: 98% CAS No.: 35704-19-9 Molecular Formula: C9H8N2O Formula Weight: 160.18 Melting Point: 204-206° MDL No.: MFCD00001814

Vzorec: C₉H₈N₂O
MW: 160.18 g/mol
Bod tání: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00001814
CAS číslo: 35704-19-9
EINECS: 252-685-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR