D-(-)-Penicillamine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
P4875-1G
P4875-25G
P4875-5G
SIALP4875-1GEA
2430
CZK
SIALP4875-1G
SIALP4875-25G
SIALP4875-5G
D-(-)-Penicillamine, Sigma-Aldrich®
D-(-)-Penicillamine
Vzorec:
C₅H₁₁NO₂S |
MDL Number:
MFCD00064302 CAS číslo: 52-67-5 EINECS: 200-148-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...