Tisk…
Phytol (mixture of cis and trans isomers), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: (7R,11R,E)-3,7,11,15-Tetramethylhexadec-2-en-1-ol
M-Clarity™ quality level = MQ300
W502200-100G W502200-1KG W502200-4KG W502200-SAMPLE
SIALW502200-100GEA 3410 CZK
SIALW502200-100G SIALW502200-1KG SIALW502200-4KG SIALW502200-SAMPLE
Phytol (mixture of cis and trans isomers), Sigma-Aldrich®
Phytol (mixture of cis and trans isomers)
Vzorec: C₂₀H₄₀O
MDL Number: MFCD00151280
CAS číslo: 150-86-7
EINECS: 205-776-6