2,4-Dimethyl-5-thiazolecarboxylic acid
Dodavatel: Cayman Chemical
Synonyms:
2,4-Dimethylthiazole-5-carboxylic acid
13233-100
13233-250
13233-500
13233-1
CAYM13233-100EA
675
CZK
CAYM13233-100
CAYM13233-250
CAYM13233-500
CAYM13233-1
2,4-Dimethyl-5-thiazolecarboxylic acid
2,4-Dimethyl-5-thiazolecarboxylic acid
Vzorec:
C₆H₇NO₂S MW: 157,1900 g/mol |
MDL Number:
MFCD00052943 CAS číslo: 53137-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...