Ethyl-2,4-dimethylthiazole-5-carboxylate
Dodavatel: Cayman Chemical
Synonyms:
Ethyl 2,4-dimethylthiazole-5-carboxylate
13232-100
13232-250
13232-500
13232-1
CAYM13232-100EA
675
CZK
CAYM13232-100
CAYM13232-250
CAYM13232-500
CAYM13232-1
Ethyl-2,4-dimethylthiazole-5-carboxylate
Ethyl-2,4-dimethylthiazole-5-carboxylate
Vzorec:
C₈H₁₁NO₂S MW: 185,24 g/mol |
MDL Number:
MFCD00052874 CAS číslo: 7210-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...