2-Acetamidot-4-methylhiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-(4-Methylthiazol-2-yl)acetamide
M-Clarity™ quality level = MQ100
301922-25G
301922-5G
SIAL301922-25GEA
12350
CZK
SIAL301922-25G
SIAL301922-5G
2-Acetamidot-4-methylhiazole, Sigma-Aldrich®
2-Acetamidot-4-methylhiazole
Vzorec:
C₆H₈N₂OS MW: 156,2055 g/mol |
MDL Number:
MFCD00011561 CAS číslo: 7336-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...