Tisk…

3-Amino-5-hydroxy-1H-pyrazole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

M-Clarity™ quality level = MQ100

331449-25G 331449-5G
SIAL331449-25GEA 2940 CZK
SIAL331449-25G SIAL331449-5G
3-Amino-5-hydroxy-1H-pyrazole, Sigma-Aldrich®
3-Amino-5-hydroxy-1H-pyrazole
Vzorec: C₃H₅N₃O
MW: 99,09 g/mol
MDL Number: MFCD00022384
CAS číslo: 6126-22-3
EINECS: 228-095-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR