Tisk…

3-Amino-5-hydroxy-1H-pyrazole 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

A11412.06 A11412.14 A11412.22
A11412.06EA 1270 CZK
A11412.06 A11412.14 A11412.22
3-Amino-5-hydroxy-1H-pyrazole 98%
3-Amino-5-hydroxy-1H-pyrazole

Purity: 98% CAS No.: 6126-22-3 Molecular Formula: C3H5N3O Formula Weight: 99.09 Odour: Odourless Melting Point: ca 210° dec. Beilstein Reference: 507793 MDL No.: MFCD00022384

Vzorec: C₃H₅N₃O
MW: 99,09 g/mol
Bod tání: ∼ 210 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00022384
CAS číslo: 6126-22-3
EINECS: 228-095-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR