Tisk…

4-Iodo-1H-pyrazole ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Iodopyrazole

H56369.06 H56369.14
H56369.06EA 3570 CZK
H56369.06 H56369.14
4-Iodo-1H-pyrazole ≥99%
4-Iodo-1H-pyrazole
Vzorec: C₃H₃IN₂
MW: 193,97 g/mol
Bod tání: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS číslo: 3469-69-0
EINECS: 222-434-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR