Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Dodavatel: Thermo Scientific
Synonyms:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
A10882.14
A10882.22
A10882.06
A10882.14EA
8410
CZK
A10882.14
A10882.22
A10882.06
Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Ethyl-3-amino-1H-pyrazole-4-carboxylate
Purity: 99%
CAS No.: 6994-25-8
Molecular Formula: C6H9N3O2
Formula Weight: 155.16
Melting Point: 104-107°
Beilstein Reference: 4964
Vzorec:
C₆H₉N₃O₂ MW: 155,16 g/mol Bod tání: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005238 CAS číslo: 6994-25-8 EINECS: 230-262-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...