Diphenylphosphine oxide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B21101.03
B21101.06
B21101.14
B21101.03EA
2170
CZK
B21101.03
B21101.06
B21101.14
Diphenylphosphine oxide ≥97%
Diphenylphosphine oxide
Purity: 97%
CAS No.: 4559-70-0
Molecular Formula: C12H11OP
Formula Weight: 202.20
Melting Point: 51-56°
MDL No.: MFCD00002079
Vzorec:
C₁₂H₁₁OP Bod tání: 51…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002079 CAS číslo: 4559-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...