Tisk…

Phenylphosphinic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

P28808-100G P28808-500G
SIALP28808-100GEA 1610 CZK
SIALP28808-100G SIALP28808-500G
Phenylphosphinic acid, Sigma-Aldrich®
Phenylphosphinic acid
Vzorec: C₆H₇O₂P
MW: 142,092 g/mol
MDL Number: MFCD00002131
CAS číslo: 1779-48-2
EINECS: 217-217-3
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR