Tisk…

Dimethylethoxysilane ≥94% ammonia-free

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L16476.14 L16476.22
L16476.14EA 2760 CZK
L16476.14 L16476.22
Dimethylethoxysilane ≥94% ammonia-free
Dimethylethoxysilane

Purity: 94% CAS No.: 14857-34-2 Molecular Formula: C4H12OSi Formula Weight: 104.23 Flash Point: 15°(59°F) Refractive Index: 1.3680 Boiling Point: 55° Beilstein Reference: 1731481 UN No.: 1993 MDL No.: MFCD00026750

Vzorec: C₄H₁₂OSi
MW: 104.23 g/mol
Bod varu: 55 °C
Hustota: 0.757
Bod vzplanutí: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026750
CAS číslo: 14857-34-2
EINECS: 238-921-7
UN: 1993
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR