9,10-Anthraquinone-2-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
252727-1G
252727-5G
SIAL252727-1GEA
1890
CZK
SIAL252727-1G
SIAL252727-5G
9,10-Anthraquinone-2-carboxylic acid, Sigma-Aldrich®
9,10-Anthraquinone-2-carboxylic acid
Vzorec:
C₁₅H₈O₄ |
MDL Number:
MFCD00001231 CAS číslo: 117-78-2 EINECS: 204-207-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...