Tisk…

4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid

Dodavatel: Cayman Chemical

Synonyms: 4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid

13036-100 13036-250 13036-500 13036-1
CAYM13036-100EA 675 CZK
CAYM13036-100 CAYM13036-250 CAYM13036-500 CAYM13036-1
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid
4-Methyl-2-(3-pyridinyl)-5-thiazolecarboxylic acid
Vzorec: C₁₀H₈N₂O₂S
MDL Number: MFCD00171753
CAS číslo: 39091-01-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR