Tisk…

2-Methyl-4-phenylthiazol

Dodavatel: Cayman Chemical

13386-100 13386-250 13386-500 13386-1
CAYM13386-100EA 1840 CZK
CAYM13386-100 CAYM13386-250 CAYM13386-500 CAYM13386-1
2-Methyl-4-phenylthiazol
2-Methyl-4-phenylthiazol
Vzorec: C₁₀H₉NS
MDL Number: MFCD00489998
CAS číslo: 1826-16-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR