Tisk…

7-Oxabicyclo[2.2.1]heptane ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

B22612.06 B22612.14 B22612.22
B22612.06EA 2810 CZK
B22612.06 B22612.14 B22612.22
7-Oxabicyclo[2.2.1]heptane ≥98%
7-Oxabicyclo[2.2.1]heptane

Purity: 98% CAS No.: 279-49-2 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 12°(55°F) Refractive Index: 1.4480 Boiling Point: 118-120°/713mm UN No.: 3271 MDL No.: MFCD00082342

Vzorec: C₆H₁₀O
MW: 98,15 g/mol
Bod varu: 118…120 °C (713 mmHg)
Hustota: 0,968
Bod vzplanutí: 12 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082342
CAS číslo: 279-49-2
EINECS: 205-998-3
UN: 3271
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR