Tisk…

4-Chloro-1,8-naphthalic anhydride ≥94%

Dodavatel: Thermo Scientific
Varování

L05508.14 L05508.22 L05508.36
L05508.14EA 1150 CZK
L05508.14 L05508.22 L05508.36
4-Chloro-1,8-naphthalic anhydride ≥94%
4-Chloro-1,8-naphthalic anhydride

Purity: 94% CAS No.: 4053-08-1 Molecular Formula: C12H5ClO3 Formula Weight: 232.63 Melting Point: 208-214° Beilstein Reference: 178072

Vzorec: C₁₂H₅ClO₃
MW: 232,63 g/mol
Bod tání: 208…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00006928
CAS číslo: 4053-08-1
EINECS: 223-760-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR