Isosorbide ≥98%
Dodavatel: Thermo Fisher Scientific
A13989.0B
A13989.18
A13989.30
A13989.0BEA
17510
CZK
A13989.0B
A13989.18
A13989.30
Isosorbide ≥98%
Isosorbide
Purity: 98%
CAS No.: 652-67-5
Molecular Formula: C6H10O4
Formula Weight: 146.14
Odour: Odourless
Melting Point: 56-64°
Beilstein Reference: 80510
Merck Reference: 14,5224
Optical Rotation: +45° (c=3 in water)
MDL No.: MFCD00064827
Vzorec:
C₆H₁₀O₄ Bod tání: 56…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064827 CAS číslo: 652-67-5 EINECS: 211-492-3 Merck Index: 14,5224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...