Tisk…

9-Phenylcarbazole ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N-Phenylcarbazole

H56827.06 H56827.14
H56827.06EA 5740 CZK
H56827.06 H56827.14
9-Phenylcarbazole ≥99%
9-Phenylcarbazole

Purity: 99% CAS No.: 1150-62-5 Molecular Formula: C18H13N Formula Weight: 243.30 Melting Point: 95-97° MDL No.: MFCD00004965

Vzorec: C₁₈H₁₃N
Bod tání: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004965
CAS číslo: 1150-62-5
EINECS: 214-564-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR