Tisk…

2,1,3-Benzothiadiazol-4-ylamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole

L09678.03 L09678.06
L09678.03EA 1940 CZK
L09678.03 L09678.06
2,1,3-Benzothiadiazol-4-ylamine 98%
2,1,3-Benzothiadiazol-4-ylamine

Purity: 98% CAS No.: 767-64-6 Molecular Formula: C6H5N3S Formula Weight: 151.19 Melting Point: 67-69° Beilstein Reference: 3551

Vzorec: C₆H₅N₃S
MW: 151,19 g/mol
Bod tání: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS číslo: 767-64-6
EINECS: 212-186-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR