3-Ethylbenzothiazolium bromide ≥98%
Dodavatel: Thermo Scientific
Synonyms:
3-Ethylbenzo[d]thiazol-3-ium bromide
L08903.09
L08903.18
L08903.09EA
2130
CZK
L08903.09
L08903.18
3-Ethylbenzothiazolium bromide ≥98%
3-Ethylbenzothiazolium bromide
Purity: 98%
CAS No.: 32446-47-2
Molecular Formula: C9H10BrNS
Formula Weight: 244.16
Melting Point: 205-207°
Beilstein Reference: 6250086
Vzorec:
C₉H₁₀BrNS MW: 244,16 g/mol Bod tání: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051852 CAS číslo: 32446-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...