Tisk…

Fmoc-L-phenylalanine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

338338-25G 338338-5G
SIAL338338-25GEA 1540 CZK
SIAL338338-25G SIAL338338-5G
Fmoc-L-phenylalanine, Sigma-Aldrich®
Fmoc-L-phenylalanine
Vzorec: C₂₄H₂₁NO₄
MDL Number: MFCD00037128
CAS číslo: 35661-40-6
EINECS: 252-661-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR