Tisk…

(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

M-Clarity™ quality level = MQ100

331910-1G 331910-5G
SIAL331910-1GEA 993 CZK
SIAL331910-1G SIAL331910-5G
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, Sigma-Aldrich®
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Vzorec: C₆H₇NO
MW: 109,1259 g/mol
MDL Number: MFCD00213364
CAS číslo: 49805-30-3
EINECS: 421-830-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR