Tisk…
2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium
A16922.AE A16922.AP A16922.0F
A16922.AEEA 1370 CZK
A16922.AE A16922.AP A16922.0F
2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
2-Mercaptopyridine-N-oxide sodium salt
CAS No.: 3811-73-2 Molecular Formula: C5H4NNaOS Formula Weight: 149.15 Refractive Index: 1.4825 Beilstein Reference: 4026050 Merck Reference: 14,7994 MDL No.: MFCD01941547
Vzorec: C₅H₄NNaOS
MW: 149.15 g/mol
Hustota: 1.238
Storage Temperature: Ambient
MDL Number: MFCD01941547
CAS číslo: 3811-73-2
EINECS: 223-296-5
Merck Index: 14,7994