Tisk…

Lead(II) acetylacetonate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Lead(II) 2,4-pentanedionate

89858.09 89858.18
89858.09EA 3850 CZK
89858.09 89858.18
Lead(II) acetylacetonate
Lead(II) acetylacetonate

CAS No.: 15282-88-9 Molecular Formula: C10H14O4Pb Formula Weight: 405.41 Physical Form: Crystalline Sensitivity: Hygroscopic Odour: Odourless UN No.: 3467 MDL No.: MFCD00013499

Vzorec: C₁₀H₁₄O₄Pb
MW: 405.41 g/mol
Bod tání: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00013499
CAS číslo: 15282-88-9
EINECS: 239-323-9
UN: 3467
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR