Tisk…

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), Ni(TMHD)2

40470.03 40470.06 40470.14
40470.03EA 3150 CZK
40470.03 40470.06 40470.14
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II)

Purity: 98% CAS No.: 14481-08-4 Molecular Formula: C22H38NiO4 Formula Weight: 425.24 Physical Form: Powder Melting Point: 223-225° Boiling Point: 90-110°/0.1mm subl. MDL No.: MFCD00192348


NMR shift reagent, nickel oxide precursor

Vzorec: C₂₂H₃₈NiO₄
MW: 425.23 g/mol
Bod varu: 90…110 °C (0.1 mmHg) (subl.)
Bod tání: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00192348
CAS číslo: 14481-08-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR