Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV) ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
Synonyms:
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), Zr(TMHD)4
Purity: 99.99%
CAS No.: 18865-74-2
Molecular Formula: C44H76)8Zr
Formula Weight: 824.30
Physical Form: Crystalline
Melting Point: 318-320°
Boiling Point: 180°/0.1mm subl.
MDL No.: MFCD00145380
MOCVD of zirconium
Vzorec:
C₄₄H₇₆O₈Zr MW: 824,29 g/mol Bod varu: 180 °C (0,1 mmHg) (subl.) Bod tání: 318…320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145380 CAS číslo: 18865-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...