Tisk…

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III) ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), Tb(TMHD)3

44842.03 44842.06
44842.03EA 3540 CZK
44842.03 44842.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')terbium(III)

Purity: 99% CAS No.: 15492-51-0 Molecular Formula: C33H57O6Tb Formula Weight: 708.74 Physical Form: Granular Melting Point: 155-156° MDL No.: MFCD00064752

Vzorec: C₃₃H₅₇O₆Tb
MW: 708.727 g/mol
Bod tání: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00064752
CAS číslo: 15492-51-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR