Tisk…

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gadolinium(III)

Dodavatel: Thermo Fisher Scientific
Varování

14486.03 14486.06
14486.03EA 2510 CZK
14486.03 14486.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gadolinium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')gadolinium(III)

CAS No.: 14768-15-1 Molecular Formula: C33H57GdO6 Formula Weight: 707.07 Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 167-170° MDL No.: MFCD00010349

Vzorec: C₃₃H₅₇GdO₆
Bod tání: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00010349
CAS číslo: 14768-15-1
EINECS: 238-834-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR