Tisk…
SparkPlugs® Disposable Earplugs
Dodavatel: Moldex
7825 785001 7800 7802 7801 785002 787001
111-1174EA 2280 CZK
111-1174 111-1175 111-1171 111-1173 111-1172 MOLD785002 MOLD787001
SparkPlugs® Disposable Earplugs
Chrániče zátkové Chrániče zátkové jednorázové
SparkPlugs® earplugs are made of a soft foam which becomes firmer when rolled for insertion.
  • Tapered shape fits easily in the ear canal
  • Seals gently and snugly without pressure
  • Appealing colours
SNR: 35 dB
Certifikace: EN 352-2:2002
Informace pro objednávání: Also available in the hygienic MoldexStation dispenser system.
Upozornení: * One size fits all.