Tisk…
Columns for phase separation, CHROMABOND® PTS/PTL
Dodavatel: MACHEREY-NAGEL
730712 730714 730716 730718 730720 730722 730724 730732 730734 730736 730738 730740 730742
MANA730712EA 4730 CZK
MANA730712 MANA730714 MANA730716 MANA730718 MANA730720 MANA730722 MANA730724 MANA730732 MANA730734 MANA730736 MANA730738 MANA730740 MANA730742
Columns for phase separation, CHROMABOND® PTS/PTL
Kolony chromatografické
CHROMABOND® PTS and CHROMABOND® PTL columns are designed for the separation of a two-phase mixture without a separation funnel.
  • The special membrane stops the flow when the lower phase has passed
  • The upper phase remains in the column, thus both phases are available for further analysis
  • Columns must not be run with vacuum or pressure
PTS for solvents heavier than water, like trichloromethane and dichloromethane. PTL for solvents lighter than water, like diethyl ether and hexane.