Tisk…
Glove liners, BCR®
Dodavatel: BERKSHIRE
BGL6.200.SB BGL7.200.SB BGL7.200.XLB BGL6.200.LB BGL7.200.MB BGL7.200.LB
BERKBGL6.200.SBEA 29110 CZK
BERKBGL6.200.SB BERKBGL7.200.SB BERKBGL7.200.XLB BERKBGL6.200.LB BERKBGL7.200.MB BERKBGL7.200.LB
Glove liners, BCR®
Rukavice
These single knit glove liners are made out of high quality 100% continuous filament pre-shrunk nylon. They are lightweight, soft, and comfortable to wear. Available as half-finger and full-finger versions.
  • Protect against skin irritation caused by latex or PVC gloves
  • Constructed using special seamless techniques
  • Ambidextrous for easy stock control
  • Launderable for multiple reuse
  • Colour-coded cuffs to identify sizes
Suitable for use in cleanrooms.