MasterSense™ MP30 Pressure Sensor with ¹/₄" NPT Process Connection
Dodavatel: Avantor Fluid Handling
High accuracy pressure transducers for plug and play connectivity with Masterflex® MasterSense™ pump systems.
- Add critical pressure monitoring to your workflow
- All-stainless welded body and wetted parts
- Rugged design for shock protection and excellent long-term stability
- CE certificated
- Sanitary models are designed for clean-in-place and sterilize-in-place
- 0 to 30 psi
- 4 to 20 mA output
- CE Compliant
- IP 65
asterSense™ MP30 pressure transducers utilize a thin-film-on-steel measurement principle to provide an accurate 0 to 30 psi output directly to MasterSense™ pump systems. Sensors include a six foot cable with AUX plug adaptor for easy plug and play.
Designed for use with a ¹/₄" FNPT process tee connection (not included)
Install downstream of pump to detect pressure changes in your process. Perfect for tangential-Flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications. Choose the model with the appropriate process connection for your application.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path. Model MFLX07526-81 ³/₄" sanitary connection not recommended for applications with high temperature variations.
<b>Certifikace: </b>CE Certified.