CPC® High-Flow Plastic Quick-Disconnect Fitting, Hose Barb Insert, Straight-Through, Polysulfone, 1/2" ID, HFC22839M; 10/PK
Dodavatel: Avantor Fluid Handling
Fittings connect in a snap.
- Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
- Fitting body constructed of durable polysulfone
These CPC® quick-disconnect fittings provide aseptic disconnect functionality. The thumb latch enables easy one-handed operation. An audible click is heard when the body and insert are coupled to confirm a secure, leak-free connection. These fittings are made of USP Class VI-compliant and BSE/TSE-free polysulfone, ideal for rugged applications that require material certification.<br /><br />For a complete fitting, order both a body and insert (order separately).