Tisk…
Masterflex® Close-Coupled Industrial Gear Pump Head with Relief Valve, Bronze; 2.1 GPM
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX70738-12
Your Organization's Price:
Rugged corrosion-resistant bronze construction provides long service life.
  • Easy assembly to NEMA and IEC motor frames
  • Maximum flow rate of 2.1 GPM
These gear pump heads are suitable for all liquids that are compatible with bronze—most common liquids are water, oil, and mild chemicals (pH from 4 to 11).<br />Choose a gear pump head to mount to your own motor (adapters for NEMA or IEC motors are sold separately below) or select a complete system with the pump head conveniently mounted to a standard (NEMA 56C) TEFC motor.