Tisk…
Masterflex® Two-Stage High-Pressure Centrifugal Pump, 63 GPM/225 ft, 304 SS; 230/460 VAC
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX75506-70
Your Organization's Price:
Two-stage design gives superior output pressures.
  • Close-coupled design saves space; simplifies maintenance and installation
  • Fabricated from 304 stainless steel for corrosion resistance
  • Back pullout design allows servicing without disturbing piping.
  • Maximum flow rate 63 GPM
  • Maximum feet of head 225 ft
  • Operates on 230/460 VAC
Pumps are non-overloading and rated for continuous duty throughout entire operating range for reliable performance and long life. Install either vertically or horizontally.