Tisk…
Masterflex® Mechanically Coupled Centrifugal Pump, 37 GPM/145 ft, 304 SS, 230/460 VAC
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX70725-06
Your Organization's Price:
Chemical-resistance pump heads provide dependable performance and continuous service.
  • Mechanically close-coupled design saves space and simplifies installation and maintenance
  • Back pullout construction — assembly and overhaul of impeller and seal without distorting suction and discharge connections
  • Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
  • Maximum flow rate 37 GPM
  • Maximum feet of head 145 ft
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.
<b>Informace o dodávce: </b>Includes conduit box for hard-wiring.