Tisk…
Masterflex® Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Brass Fittings with valve, 22 L/min Nitrogen
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX68560-75
Your Organization's Price:
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
  • Flowmeter features brass fittings and Viton® o-ring
Versatile acrylic flowmeters are designed specifically for nitrogen. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
<B>Warning:</B> This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.