Tisk…
Hose Fittings, Band Clamps, for 2" OD Hose, 25/Pk
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX33350-14
Your Organization's Price:
Quick and easy assembly without tools.
  • Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.