Masterflex® Low Pressure-Drop Flow Switches for Liquids, Closed; 5.0 GPM
Dodavatel: Avantor Fluid Handling
Minimize flow constriction and achieve low pressure drop.
- Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 Stainless Steel, and polypropylene
- Effortlessly install with 1/2" NPT(M) connections
- Durable switch with 212 °F (100 °C) max temperature and 200 psi at 70 °F max pressure
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 µm filter to protect your switch. Switches include 1/4" male quick connect terminals.