Tisk…
Micropump L24566 Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX07003-96
Your Organization's Price:
Easily adapt your pump head to your drive with these adapter kits.
  • Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.