Tisk…
LMI RPM 312/318 Service Kit for Solenoid Diaphragm Metering Pumps, 8.0 GPH
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX74513-48
Your Organization's Price:
Optimize performance of your LMI Solenoid Diaphragm Metering Pumps.
  • Keep your pump running smoothly with these service kits
  • This kit is for : 812XX, 818XX, 910NX, 910SX
Replace wearable parts as part of a regular maintenance program to prevent failures and downtime.
<B>Service kits include:</B> a replacement diaphragm, seal, four pump head screws, and check valves.