Tisk…
High-Head Enclosed Motor PP Centrifugal Pump; 11.4 GPM/43.2 ft, 230V
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX72012-25
Your Organization's Price:
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
<b>Informace o dodávce: </b>Includes a 6-ft (1.8 m) cord with stripped ends.