Tisk…
Sanitary In-Line Strainer Assembly; 2" ; with 1/4" Perforated Core
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX29590-14
Your Organization's Price:
Protect products and expensive process equipment from coarse or fine particulates.
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
<b>Informace o dodávce: </b>Strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.